Assay Details
Amplicon Length: | 70 |
Chromosome Location: | Chr. 5 - 138166133 - 138166133 [+] on 138166133 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-106b-3p | MIMAT0004582 |
Mouse | mmu-miR-106b-5p | MIMAT0000386 |
Stem-loop Details
Mouse
