Assay Details
Amplicon Length: | 65 |
Chromosome Location: | Chr. 7 - 100094396 - 100094396 [+] on 100094396 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-106b-3p | MIMAT0004672 |
| Human | hsa-miR-106b-5p | MIMAT0000680 |
Stem-loop Details
Human

Genomic Map