Assay Details
Amplicon Length: | 63 |
Chromosome Location: | Chr. 12 - 19308105 - 19308105 [-] on 19308105 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-106b-3p | MIMAT0004727 |
Rat | rno-miR-106b-5p | MIMAT0000825 |
Stem-loop Details
Rat
