Assay Details
Gene Family ID: | MIPF0000130, mir-506 |
Mature miRNA Sequence: | UAAGGCACCCUUCUGAGUAGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-506-3p | MIMAT0002878 |
| Pan troglodytes | ptr-miR-506 | MIMAT0005770 |
| Pongo pygmaeus | ppy-miR-506 | MIMAT0015929 |
| Pygathrix bieti | pbi-miR-506 | MIMAT0005779 |
| Rhesus monkey | mml-miR-506-3p | MIMAT0005762 |
| Symphalangus syndactylus | ssy-miR-506 | MIMAT0005752 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
Pygathrix bieti
Rhesus monkey
Symphalangus syndactylus

Genomic Map