Assay Details
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | CUAGACUGAGGCUCCUUGAGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-151-3p | MIMAT0000161 |
Rat | rno-miR-151-3p | MIMAT0000614 |
Cricetulus griseus | cgr-miR-151-3p | MIMAT0023783 |
Guinea pig | cpo-miR-151-3p | MIMAT0047031 |
Microcebus murinus | mmr-miR-151 | MIMAT0049221 |
Stem-loop Details
Mouse
Rat
Cricetulus griseus
Guinea pig
Microcebus murinus

More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-151 (v9.2)
Rat: rno-miR-151* (v18)
Rat: rno-miR-151* (v18)
