Assay Details
Amplicon Length: | 66 |
Chromosome Location: | Chr. 15 - 73255018 - 73255018 [+] on 73255018 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-151-5p | MIMAT0004536 |
| Mouse | mmu-miR-151-3p | MIMAT0000161 |
Stem-loop Details
Mouse

Genomic Map