Assay Details
| Gene Family ID: | MIPF0000057, mir-28 | 
| Mature miRNA Sequence: | CUAGACUGAGGCUCCUUGAGG | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Mouse | mmu-miR-151-3p | MIMAT0000161 | 
| Rat | rno-miR-151-3p | MIMAT0000614 | 
| Cricetulus griseus | cgr-miR-151-3p | MIMAT0023783 | 
| Guinea pig | cpo-miR-151-3p | MIMAT0047031 | 
| Microcebus murinus | mmr-miR-151 | MIMAT0049221 | 
Stem-loop Details
Mouse
Rat
Cricetulus griseus
Guinea pig
Microcebus murinus
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top TaqMan™ Pri-miRNA Assay :
						TaqMan™ Pri-miRNA Assay :