Assay Details
Amplicon Length: | 64 |
Chromosome Location: | Chr. 7 - 114485992 - 114485992 [-] on 114485992 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-151-5p | MIMAT0000613 |
Rat | rno-miR-151-3p | MIMAT0000614 |
Rat | rno-miR-3586-5p | MIMAT0017880 |
Rat | rno-miR-3586-3p | MIMAT0017881 |
Stem-loop Details
Rat
