Assay Details
Gene Family ID: | MIPF0000183, mir-503 |
Mature miRNA Sequence: | UAGCAGCGGGAACAGUUCUGCAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-503-5p | MIMAT0002874 |
Gorilla gorilla | ggo-miR-503 | MIMAT0024203 |
Pan troglodytes | ptr-miR-503 | MIMAT0008176 |
Pongo pygmaeus | ppy-miR-503 | MIMAT0015926 |
Rhesus monkey | mml-miR-503-5p | MIMAT0006367 |
Stem-loop Details
Human
Gorilla gorilla
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

More Information
Related Products
miRBase Alias:
Human: hsa-miR-503 (v18)
Macaca mulatta: mml-miR-503 (v19)
Macaca mulatta: mml-miR-503 (v19)
