Assay Details
Gene Family ID: | MIPF0000084, mir-142 |
Mature miRNA Sequence: | CAUAAAGUAGAAAGCACUAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-142-5p | MIMAT0003790 |
| Xenopus laevis | xla-miR-142 | MIMAT0011139 |
| Xenopus tropicalis | xtr-miR-142-5p | MIMAT0003602 |
Stem-loop Details
Bovine
Xenopus laevis
Xenopus tropicalis
More Information
Related Products
miRBase Alias:
Human: hsa-miR-142-5p (v9.2)
Mouse: mmu-miR-142-5p (v9.2)
Rat: rno-miR-142-5p (v9.2)
Bos taurus: bta-miR-142 (v18)
Gallus gallus: gga-miR-142-5p (v8.1)
Mouse: mmu-miR-142-5p (v9.2)
Rat: rno-miR-142-5p (v9.2)
Bos taurus: bta-miR-142 (v18)
Gallus gallus: gga-miR-142-5p (v8.1)

Genomic Map