Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | CACACACUGCAAUUACUUUUGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-603 | MIMAT0003271 |
| Pongo pygmaeus | ppy-miR-603 | MIMAT0016045 |
Stem-loop Details
Human
Pongo pygmaeus

Genomic Map