Assay Details
Gene Family ID: | MIPF0000109, mir-186 |
Mature miRNA Sequence: | GCCCAAAGGUGAAUUUUUUGGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-186-3p | MIMAT0004612 |
| Dasypus novemcinctus | dno-miR-186-3p | MIMAT0047708 |
| Guinea pig | cpo-miR-186-3p | MIMAT0047057 |
| Pig | ssc-miR-186-3p | MIMAT0037321 |
| Rabbit | ocu-miR-186-3p | MIMAT0048294 |
Stem-loop Details
Human
Dasypus novemcinctus
Guinea pig
Pig
Rabbit

Genomic Map