Assay Details
Gene Family ID: | MIPF0000180, mir-483 |
Mature miRNA Sequence: | UCACUCCUCUCCUCCCGUCUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-483-3p | MIMAT0002173 |
| Bovine | bta-miR-483 | MIMAT0009327 |
Stem-loop Details
Human
Bovine

Genomic Map