Assay Details
Gene Family ID: | MIPF0000130, mir-506 |
Mature miRNA Sequence: | AUGCCUUUUGCUCUGCACUCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-511-5p | MIMAT0004940 |
Stem-loop Details
Mouse
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-511 (v15)
Rat: rno-miR-511 (v15)
Rat: rno-miR-511 (v15)

Genomic Map