Assay Details
Gene Family ID: | MIPF0000034, mir-130 |
Mature miRNA Sequence: | GCUCUGACUUUAUUGCACUACU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-301a-5p | MIMAT0022696 |
| Mouse | mmu-miR-301a-5p | MIMAT0017008 |
| Alligator mississippiensis | ami-miR-301a-5p | MIMAT0038260 |
| Chicken | gga-miR-301b-5p | MIMAT0007741 |
| Columba livia | cli-miR-301a-5p | MIMAT0038611 |
| Cricetulus griseus | cgr-miR-301a-5p | MIMAT0023890 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-301a-5p | MIMAT0047787 |
| Guinea pig | cpo-miR-301a-5p | MIMAT0047142 |
| Pteropus alecto | pal-miR-301a-5p | MIMAT0040045 |
| Rabbit | ocu-miR-301a-5p | MIMAT0048364 |
| Taeniopygia guttata | tgu-miR-301-5p | MIMAT0017391 |
| Xenopus laevis | xla-miR-301-5p | MIMAT0046566 |
Stem-loop Details
Human
Mouse
Alligator mississippiensis
Chicken
Columba livia
Cricetulus griseus
Dasypus novemcinctus
Guinea pig
Pteropus alecto
Rabbit
Taeniopygia guttata
Xenopus laevis

Genomic Map