Assay Details
Gene Family ID: | MIPF0001578, mir-4774 |
Mature miRNA Sequence: | UCUGGUAUGUAGUAGGUAAUAA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-4774-5p | MIMAT0019929 |
| Pongo pygmaeus | ppy-miR-4774 | MIMAT0024382 |
Stem-loop Details
Human
Pongo pygmaeus

Genomic Map