Assay Details
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | UAAAGUGCUGACAGUGCAGAU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-106b-5p | MIMAT0000680 |
Mouse | mmu-miR-106b-5p | MIMAT0000386 |
Rat | rno-miR-106b-5p | MIMAT0000825 |
Ateles geoffroyi | age-miR-106b | MIMAT0002761 |
Bovine | bta-miR-106b | MIMAT0009218 |
Callithrix jacchus | cja-miR-106b | MIMAT0049318 |
View More Rows | ||
Dog | cfa-miR-106b | MIMAT0006695 |
Goat | chi-miR-106b-5p | MIMAT0035908 |
Gorilla gorilla | ggo-miR-106b | MIMAT0002758 |
Horse | eca-miR-106b | MIMAT0013054 |
Lagothrix lagotricha | lla-miR-106b | MIMAT0002777 |
Macaca nemestrina | mne-miR-106b | MIMAT0002780 |
Pan paniscus | ppa-miR-106b | MIMAT0002763 |
Pan troglodytes | ptr-miR-106b | MIMAT0002769 |
Pongo pygmaeus | ppy-miR-106b | MIMAT0002766 |
Rhesus monkey | mml-miR-106b-5p | MIMAT0002772 |
Saguinus labiatus | sla-miR-106b | MIMAT0002775 |
Sheep | oar-miR-106b | MIMAT0030028 |
Stem-loop Details
Human
Mouse
Rat
Ateles geoffroyi
Bovine
Callithrix jacchus
Dog
Goat
Gorilla gorilla
Horse
Lagothrix lagotricha
Macaca nemestrina
Pan paniscus
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
Saguinus labiatus
Sheep
