Assay Details
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | UAAAGUGCUGACAGUGCAGAU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-106b-5p | MIMAT0000680 | |
| Mouse | mmu-miR-106b-5p | MIMAT0000386 | |
| Rat | rno-miR-106b-5p | MIMAT0000825 | |
| Ateles geoffroyi | age-miR-106b | MIMAT0002761 | |
| Bovine | bta-miR-106b | MIMAT0009218 | |
| Callithrix jacchus | cja-miR-106b | MIMAT0049318 | |
| View More Rows | |||
| Dog | cfa-miR-106b | MIMAT0006695 | |
| Goat | chi-miR-106b-5p | MIMAT0035908 | |
| Gorilla gorilla | ggo-miR-106b | MIMAT0002758 | |
| Horse | eca-miR-106b | MIMAT0013054 | |
| Lagothrix lagotricha | lla-miR-106b | MIMAT0002777 | |
| Macaca nemestrina | mne-miR-106b | MIMAT0002780 | |
| Pan paniscus | ppa-miR-106b | MIMAT0002763 | |
| Pan troglodytes | ptr-miR-106b | MIMAT0002769 | |
| Pongo pygmaeus | ppy-miR-106b | MIMAT0002766 | |
| Rhesus monkey | mml-miR-106b-5p | MIMAT0002772 | |
| Saguinus labiatus | sla-miR-106b | MIMAT0002775 | |
| Sheep | oar-miR-106b | MIMAT0030028 | |
Stem-loop Details
Human
Mouse
Rat
Ateles geoffroyi
Bovine
Callithrix jacchus
Dog
Goat
Gorilla gorilla
Horse
Lagothrix lagotricha
Macaca nemestrina
Pan paniscus
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
Saguinus labiatus
Sheep
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products
miRBase Alias:
Human: hsa-miR-106b (v17)
Mouse: mmu-miR-106b (v17)
Rat: rno-miR-106b (v18)
Macaca mulatta: mml-miR-106b (v19)
Mouse: mmu-miR-106b (v17)
Rat: rno-miR-106b (v18)
Macaca mulatta: mml-miR-106b (v19)

Genomic Map