Assay Details
Amplicon Length: | 112 |
Chromosome Location: | Chr. 17 - 28861061 - 28861061 [+] on 28861061 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-451a | MIMAT0001631 |
| Human | hsa-miR-451b | MIMAT0019840 |
Stem-loop Details
Human

Genomic Map