Assay Details
Amplicon Length: | 71 |
Chromosome Location: | Chr. X - 74217976 - 74217976 [-] on 74217976 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-421 | MIMAT0003339 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 71 |
Chromosome Location: | Chr. X - 74217976 - 74217976 [-] on 74217976 Build GRCh38 |
Species: | Human |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-421 | MIMAT0003339 |