Assay Details
| Gene Family ID: | MIPF0000098, mir-95 | 
| Mature miRNA Sequence: | AUCAACAGACAUUAAUUGGGCGC | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-421 | MIMAT0003339 | 
| Mouse | mmu-miR-421-3p | MIMAT0004869 | 
| Bovine | bta-miR-421 | MIMAT0009314 | 
| Callithrix jacchus | cja-miR-421 | MIMAT0039378 | 
| Goat | chi-miR-421-3p | MIMAT0036219 | 
| Nomascus leucogenys | nle-miR-421 | MIMAT0049415 | 
| View More Rows | ||
| Pan troglodytes | ptr-miR-421 | MIMAT0008140 | 
| Pig | ssc-miR-421-3p | MIMAT0022962 | 
| Pongo pygmaeus | ppy-miR-421 | MIMAT0015880 | 
| Rabbit | ocu-miR-421-3p | MIMAT0048440 | 
| Rhesus monkey | mml-miR-421 | MIMAT0006319 | 
Stem-loop Details
Human
Mouse
Bovine
Callithrix jacchus
Goat
Nomascus leucogenys
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top TaqMan™ Pri-miRNA Assay :
						TaqMan™ Pri-miRNA Assay :