Assay Details
Gene Family ID: | MIPF0000098, mir-95 |
Mature miRNA Sequence: | AUCAACAGACAUUAAUUGGGCGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-421 | MIMAT0003339 | |
| Mouse | mmu-miR-421-3p | MIMAT0004869 | |
| Bovine | bta-miR-421 | MIMAT0009314 | |
| Callithrix jacchus | cja-miR-421 | MIMAT0039378 | |
| Goat | chi-miR-421-3p | MIMAT0036219 | |
| Nomascus leucogenys | nle-miR-421 | MIMAT0049415 | |
| View More Rows | |||
| Pan troglodytes | ptr-miR-421 | MIMAT0008140 | |
| Pig | ssc-miR-421-3p | MIMAT0022962 | |
| Pongo pygmaeus | ppy-miR-421 | MIMAT0015880 | |
| Rabbit | ocu-miR-421-3p | MIMAT0048440 | |
| Rhesus monkey | mml-miR-421 | MIMAT0006319 | |
Stem-loop Details
Human
Mouse
Bovine
Callithrix jacchus
Goat
Nomascus leucogenys
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey

Genomic Map