Assay Details
Gene Family ID: | MIPF0000098, mir-95 |
Mature miRNA Sequence: | AUCAACAGACAUUAAUUGGGCGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-421 | MIMAT0003339 |
Mouse | mmu-miR-421-3p | MIMAT0004869 |
Bovine | bta-miR-421 | MIMAT0009314 |
Callithrix jacchus | cja-miR-421 | MIMAT0039378 |
Goat | chi-miR-421-3p | MIMAT0036219 |
Nomascus leucogenys | nle-miR-421 | MIMAT0049415 |
View More Rows | ||
Pan troglodytes | ptr-miR-421 | MIMAT0008140 |
Pig | ssc-miR-421-3p | MIMAT0022962 |
Pongo pygmaeus | ppy-miR-421 | MIMAT0015880 |
Rabbit | ocu-miR-421-3p | MIMAT0048440 |
Rhesus monkey | mml-miR-421 | MIMAT0006319 |
Stem-loop Details
Human
Mouse
Bovine
Callithrix jacchus
Goat
Nomascus leucogenys
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey
