Assay Details
Amplicon Length: | 76 |
Chromosome Location: | Chr. 4 - 20527978 - 20527978 [+] on 20527978 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-218-5p | MIMAT0000275 |
| Human | hsa-miR-218-1-3p | MIMAT0004565 |
Stem-loop Details
Human

Genomic Map