Assay Details
Gene Family ID: | MIPF0000186, mir-299 |
Mature miRNA Sequence: | UGGUUUACCGUCCCACAUACAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-299-5p | MIMAT0002890 |
| Mouse | mmu-miR-299a-5p | MIMAT0000377 |
| Rat | rno-miR-299a-5p | MIMAT0000901 |
| Bovine | bta-miR-299 | MIMAT0009274 |
| Dog | cfa-miR-299 | MIMAT0009885 |
| Pan troglodytes | ptr-miR-299 | MIMAT0012801 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-299-5p | MIMAT0015813 |
| Pteropus alecto | pal-miR-299a-5p | MIMAT0039971 |
| Rhesus monkey | mml-miR-299-5p | MIMAT0006255 |
| Sheep | oar-miR-299-5p | MIMAT0019251 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Dog
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rhesus monkey
Sheep
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-299-5p (v18)
Mouse: mmu-miR-299* (v17)
Mouse: mmu-miR-299 (v9.2)
Rat: rno-miR-299 (v18)
Mouse: mmu-miR-299* (v17)
Mouse: mmu-miR-299 (v9.2)
Rat: rno-miR-299 (v18)

Genomic Map