Assay Details
Gene Family ID: | MIPF0000186, mir-299 |
Mature miRNA Sequence: | UGGUUUACCGUCCCACAUACAU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-299-5p | MIMAT0002890 | |
| Mouse | mmu-miR-299a-5p | MIMAT0000377 | |
| Rat | rno-miR-299a-5p | MIMAT0000901 | |
| Bovine | bta-miR-299 | MIMAT0009274 | |
| Dog | cfa-miR-299 | MIMAT0009885 | |
| Pan troglodytes | ptr-miR-299 | MIMAT0012801 | |
| View More Rows | |||
| Pongo pygmaeus | ppy-miR-299-5p | MIMAT0015813 | |
| Pteropus alecto | pal-miR-299a-5p | MIMAT0039971 | |
| Rhesus monkey | mml-miR-299-5p | MIMAT0006255 | |
| Sheep | oar-miR-299-5p | MIMAT0019251 | |
Stem-loop Details
Human
Mouse
Rat
Bovine
Dog
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rhesus monkey
Sheep
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool B v3.0
Related Products
miRBase Alias:
Mouse: mmu-miR-299-5p (v18)
Mouse: mmu-miR-299* (v17)
Mouse: mmu-miR-299 (v9.2)
Rat: rno-miR-299 (v18)
Mouse: mmu-miR-299* (v17)
Mouse: mmu-miR-299 (v9.2)
Rat: rno-miR-299 (v18)

Genomic Map