Assay Details
Amplicon Length: | 83 |
Chromosome Location: | Chr. 6 - 18572085 - 18572085 [+] on 18572085 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548a-3p | MIMAT0003251 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 83 |
Chromosome Location: | Chr. 6 - 18572085 - 18572085 [+] on 18572085 Build GRCh38 |
Species: | Human |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548a-3p | MIMAT0003251 |