Assay Details
Amplicon Length: | 64 |
Chromosome Location: | Chr. 6 - 119069450 - 119069450 [-] on 119069450 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548b-5p | MIMAT0004798 |
| Human | hsa-miR-548b-3p | MIMAT0003254 |
Stem-loop Details
Human

Genomic Map