Assay Details
Amplicon Length: | 60 |
Chromosome Location: | Chr. 6 - 135239457 - 135239457 [-] on 135239457 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548a-3p | MIMAT0003251 |
Stem-loop Details
Human
