Assay Details
Amplicon Length: | 122 |
Chromosome Location: | Chr. 8 - 99537133 - 99537133 [+] on 99537133 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-875-3p | MIMAT0004923 |
Human | hsa-miR-875-5p | MIMAT0004922 |
Stem-loop Details
Human
