Assay Details
Gene Family ID: | MIPF0000424, mir-875 |
Mature miRNA Sequence: | UAUACCUCAGUUUUAUCAGGUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-875-5p | MIMAT0004922 | |
| Mouse | mmu-miR-875-5p | MIMAT0004937 | |
| Rat | rno-miR-875 | MIMAT0012842 | |
| Bovine | bta-miR-875 | MIMAT0009379 | |
| Callithrix jacchus | cja-miR-875-5p | MIMAT0039607 | |
| Dog | cfa-miR-875 | MIMAT0009931 | |
| View More Rows | |||
| Pan troglodytes | ptr-miR-875 | MIMAT0008326 | |
| Pongo pygmaeus | ppy-miR-875-5p | MIMAT0016113 | |
| Rabbit | ocu-miR-875-5p | MIMAT0048577 | |
| Rhesus monkey | mml-miR-875-5p | MIMAT0006517 | |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Dog
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool B v3.0Megaplex™ RT Primers, Human Pool B v3.0
Associated TaqMan® Array MicroRNA Cards:
TaqMan® Human MicroRNA B Cards v3.0TaqMan® Rodent MicroRNA B Cards v3.0
TaqMan® OpenArray® MicroRNA Panels:
TaqMan® OpenArray® Human MicroRNA PanelTaqMan® OpenArray® Rodent MicroRNA Panel
Related Products

Genomic Map