Assay Details
Gene Family ID: | MIPF0000424, mir-875 |
Mature miRNA Sequence: | UAUACCUCAGUUUUAUCAGGUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-875-5p | MIMAT0004922 |
Mouse | mmu-miR-875-5p | MIMAT0004937 |
Rat | rno-miR-875 | MIMAT0012842 |
Bovine | bta-miR-875 | MIMAT0009379 |
Callithrix jacchus | cja-miR-875-5p | MIMAT0039607 |
Dog | cfa-miR-875 | MIMAT0009931 |
View More Rows | ||
Pan troglodytes | ptr-miR-875 | MIMAT0008326 |
Pongo pygmaeus | ppy-miR-875-5p | MIMAT0016113 |
Rabbit | ocu-miR-875-5p | MIMAT0048577 |
Rhesus monkey | mml-miR-875-5p | MIMAT0006517 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Dog
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey
