Assay Details
Gene Family ID: | MIPF0000424, mir-875 |
Mature miRNA Sequence: | UAUACCUCAGUUUUAUCAGGUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-875-5p | MIMAT0004922 |
| Mouse | mmu-miR-875-5p | MIMAT0004937 |
| Rat | rno-miR-875 | MIMAT0012842 |
| Bovine | bta-miR-875 | MIMAT0009379 |
| Callithrix jacchus | cja-miR-875-5p | MIMAT0039607 |
| Dog | cfa-miR-875 | MIMAT0009931 |
| View More Rows | ||
| Pan troglodytes | ptr-miR-875 | MIMAT0008326 |
| Pongo pygmaeus | ppy-miR-875-5p | MIMAT0016113 |
| Rabbit | ocu-miR-875-5p | MIMAT0048577 |
| Rhesus monkey | mml-miR-875-5p | MIMAT0006517 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Dog
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey

Genomic Map