Assay Details
Amplicon Length: | 147 |
Chromosome Location: | Chr. 10 - 29544806 - 29544806 [+] on 29544806 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-604 | MIMAT0003272 |
Stem-loop Details
Human
