Assay Details
Amplicon Length: | 79 |
Chromosome Location: | Chr. 20 - 63942431 - 63942431 [-] on 63942431 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-647 | MIMAT0003317 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 79 |
Chromosome Location: | Chr. 20 - 63942431 - 63942431 [-] on 63942431 Build GRCh38 |
Species: | Human |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-647 | MIMAT0003317 |