Assay Details
Amplicon Length: | 55 |
Chromosome Location: | Chr. 14 - 101031587 - 101031587 [+] on 101031587 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-543 | MIMAT0004954 |
Stem-loop Details
Human
