Assay Details
Amplicon Length: | 62 |
Chromosome Location: | Chr. 3 - 197674101 - 197674101 [-] on 197674101 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-922 | MIMAT0004972 |
Stem-loop Details
Human
