Assay Details
Amplicon Length: | 87 |
Chromosome Location: | Chr. 5 - 100816188 - 100816188 [+] on 100816188 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548p | MIMAT0005934 |
Stem-loop Details
Human
