Assay Details
Amplicon Length: | 102 |
Chromosome Location: | Chr. 9 - 128245199 - 128245199 [+] on 128245199 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-199b-3p | MIMAT0004563 |
| Human | hsa-miR-199b-5p | MIMAT0000263 |
| Human | hsa-miR-3154 | MIMAT0015028 |
Stem-loop Details
Human

Genomic Map