Assay Details
Amplicon Length: | 86 |
Chromosome Location: | Chr. 1 - 22719373 - 22719373 [-] on 22719373 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4684-3p | MIMAT0019770 |
Human | hsa-miR-4684-5p | MIMAT0019769 |
Stem-loop Details
Human
