Assay Details
Amplicon Length: | 93 |
Chromosome Location: | Chr. 2 - 231362956 - 231362956 [-] on 231362956 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4777-5p | MIMAT0019934 |
Human | hsa-miR-4777-3p | MIMAT0019935 |
Stem-loop Details
Human
