Assay Details
Amplicon Length: | 76 |
Chromosome Location: | Chr. 15 - 25848596 - 25848596 [+] on 25848596 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-4715-3p | MIMAT0019825 |
| Human | hsa-miR-4715-5p | MIMAT0019824 |
Stem-loop Details
Human

Genomic Map