Assay Details
Amplicon Length: | 105 |
Chromosome Location: | Chr. 17 - 44207989 - 44207989 [-] on 44207989 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6782-3p | MIMAT0027465 |
Human | hsa-miR-6782-5p | MIMAT0027464 |
Stem-loop Details
Human
