Assay Details
Amplicon Length: | 105 |
Chromosome Location: | Chr. 1 - 12578676 - 12578676 [-] on 12578676 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-6730-3p | MIMAT0027362 |
| Human | hsa-miR-6730-5p | MIMAT0027361 |
Stem-loop Details
Human

Genomic Map