Assay Details
Amplicon Length: | 117 |
Chromosome Location: | Chr. 3 - 123162111 - 123162111 [-] on 123162111 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-7110-5p | MIMAT0028117 |
Human | hsa-miR-7110-3p | MIMAT0028118 |
Stem-loop Details
Human
