Assay Details
Mature miRNA Sequence: | AUUAACGCUGGCGGUUGCGGCAGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR404 | MIMAT0001005 |
Stem-loop Details
Arabidopsis
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ath-miR404 |
|---|---|
| miRBase Accession Number: | MI0001073 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AUUAACGCUGGCGGUUGCGGCAGC |
| Species: | Arabidopsis |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | AUUAACGCUGGCGGUUGCGGCAGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR404 | MIMAT0001005 |