Assay Details
Gene Family ID: | MIPF0000111, mir-489 |
Mature miRNA Sequence: | AGUGACAUCAUAUGUACGGCUGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chicken | gga-miR-489-3p | MIMAT0003365 |
| Fugu rubripes | fru-miR-489 | MIMAT0002938 |
| Tetraodon nigroviridis | tni-miR-489 | MIMAT0002939 |
| Xenopus tropicalis | xtr-miR-489 | MIMAT0003642 |
| Zebrafish | dre-miR-489 | MIMAT0002940 |
Stem-loop Details
Chicken
Fugu rubripes
Tetraodon nigroviridis
Xenopus tropicalis
Zebrafish

Genomic Map