Assay Details
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | UUAAGACUUGCAGUGAUGUUUAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ophiophagus hannah | oha-miR-499-5p | MIMAT0036938 |
| Xenopus tropicalis | xtr-miR-499 | MIMAT0003643 |
Stem-loop Details
Ophiophagus hannah
Xenopus tropicalis

Genomic Map