Assay Details
Gene Family ID: | MIPF0000535, mir-569 |
Mature miRNA Sequence: | AGUUAAUGAAUCCUGGAAAGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-569 | MIMAT0003234 |
Pan troglodytes | ptr-miR-569 | MIMAT0008247 |
Pongo pygmaeus | ppy-miR-569 | MIMAT0016013 |
Rhesus monkey | mml-miR-569 | MIMAT0006432 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
