Assay Details
Gene Family ID: | MIPF0000162, mir-367 |
Mature miRNA Sequence: | GAAUUGCACUUUAGCAAUGGUGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-367 | MIMAT0009299 |
| Monodelphis domestica | mdo-miR-367 | MIMAT0004184 |
Stem-loop Details
Bovine
Monodelphis domestica

Genomic Map